Recombinat Protein,Human CD23 Fc epsilon RII Protein, His Tag,Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HE K293). It contains AA Asp 48 - Ser 321 (Accession P06734-1).This protein carries a polyhistidine tag at the N-terminu 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD23 Fc epsilon RII Protein, His Tag,Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HE K293). It contains AA Asp 48 - Ser 321 (Accession P06734-1).This protein carries a polyhistidine tag at the N-terminu 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD23 Fc epsilon RII Protein, His Tag,Human CD23, His Tag (CD3-H5249) is expressed from human 293 cells (HE K293). It contains AA Asp 48 - Ser 321 (Accession P06734-1).This protein carries a polyhistidine tag at the N-terminu 1 * 100 µG ACRBCD3-H5249100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou