Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human Siglec-3 CD33 (18-132) Protein, Fc Tag,Human Siglec-3 (18-132), Fc Tag (CD3-H5255) is expressed from human 293 cells (HEK293). It contains AA Pro 18 - His 132 (Accession P20138-3).This protein carries a human IgG1 Fc tag at s 1 * 100 µG ACRBCD3-H5255100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou