Recombinat Protein,Human CD163 Protein, His Tag,Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It cont ains AA Ser 42 - Ser 1045 (Accession Q86VB7).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD163 Protein, His Tag,Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It cont ains AA Ser 42 - Ser 1045 (Accession Q86VB7).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 1 mg ACRBCD3-H52E8-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou