Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD133, His Tag (CD3-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 20 - His 865 (Accession O43490-1). 1 * 100 µG ACRBCD3-H52H4100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou