Recombinat Protein,Mouse CD73 NT5E Protein, His Tag,Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 29 - Ser 551 (Accession Q61503-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 20 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse CD73 NT5E Protein, His Tag,Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 29 - Ser 551 (Accession Q61503-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 20 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse CD73 NT5E Protein, His Tag,Mouse CD73, His Tag (CD3-M52H9) is expressed from human 293 cells (HEK293). It contains AA Trp 29 - Ser 551 (Accession Q61503-1).This protein carries a polyhistidine tag at the C-terminus. The prot 1 * 20 µG ACRBCD3-M52H9-20UG 1 * 20 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou