Recombinat Protein,Rhesus macaque CD4 Protein, His Tag,Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HE K293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque CD4 Protein, His Tag,Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HE K293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Rhesus macaque CD4 Protein, His Tag,Rhesus macaque CD4, His Tag (CD4-C52H7) is expressed from human 293 cells (HE K293). It contains AA Lys 26 - Trp 390 (Accession G7N5T8).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg ACRBCD4-C52H7-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou