Human CD44, His Tag (CD4-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession AAH04372). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD44, His Tag (CD4-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession AAH04372). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD44, His Tag (CD4-H5226) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession AAH04372). 1 * 100 µG ACRBCD4-H5226100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou