Recombinat Protein,Human CD14 Protein, His Tag,Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Met 344 (Accession AAH10507).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD14 Protein, His Tag,Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Met 344 (Accession AAH10507).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD14 Protein, His Tag,Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Met 344 (Accession AAH10507).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBCD4-H5228100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou