Recombinat Protein,Human CD84 SLAMF5 Protein, His Tag,Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). I t contains AA Lys 22 - Gly 225 (Accession AAH20063).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD84 SLAMF5 Protein, His Tag,Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). I t contains AA Lys 22 - Gly 225 (Accession AAH20063).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD84 SLAMF5 Protein, His Tag,Human CD84, His Tag (CD4-H5229) is expressed from human 293 cells (HEK293). I t contains AA Lys 22 - Gly 225 (Accession AAH20063).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg ACRBCD4-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou