Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD74 DHLAG Protein, His Tag,human CD74, His Tag (CD4-H524c) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession NP004346.1).This protein carries a polyhistidine tag at the N-terminus. The ca 1 * 100 µG ACRBCD4-H524C100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou