Recombinat Protein,Human CD94 Protein, Mouse IgG2a Fc Tag,Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cel ls (HEK293). It contains AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a mouse IgG2a Fc tag at the C 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD94 Protein, Mouse IgG2a Fc Tag,Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cel ls (HEK293). It contains AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a mouse IgG2a Fc tag at the C 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD94 Protein, Mouse IgG2a Fc Tag,Human CD94, Mouse IgG2a Fc Tag (CD4-H5253) is expressed from human 293 cel ls (HEK293). It contains AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a mouse IgG2a Fc tag at the C 1 * 1 mg ACRBCD4-H5253-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou