Human CD24, His Tag (CD4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human CD24, His Tag (CD4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human CD24, His Tag (CD4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25063-1). 1 * 100 µG ACRBCD4-H52H3100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou