Recombinat Protein,Human CD94 Protein, His Tag,Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contai ns AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD94 Protein, His Tag,Human CD94, His Tag (CD4-H52H8) is expressed from human 293 cells (HEK293). It contai ns AA Lys 32 - Ile 179 (Accession Q13241-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBCD4-H52H8100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou