FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 G
COMMANDE
Développer tout / Réduire tout
FITC-Labeled Human CD4, His Tag (CD4-HF2H7) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). It is the FITC labeled form of Human CD4, His Tag (LE3-H5228). 1 * 200 µG ACRBCD4-HF2H7200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou