Mouse CD64, His Tag (CD4-M5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 297 (Accession P26151-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse CD64, His Tag (CD4-M5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 297 (Accession P26151-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse CD64, His Tag (CD4-M5227) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 297 (Accession P26151-1). 1 * 1 mg ACRBCD4-M5227-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou