Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It c ontains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pr 1 * 1 mg ACRBCD5-H5225-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou