Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus Rhesus macaque B7-2, His Tag (HPLC-verified) (CD6-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - His 240 (Accession G7NXR4-1). In the region Ala 19 - His 240, the AA sequence of Cynomolgus and Rhesus macaque B7 1 * 1 mg ACRBCD6-C52H5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou