Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession AAH40261). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession AAH40261). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession AAH40261). 1 * 1 mg ACRBCD6-H5223-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou