Recombinat Protein,Human CD46 Protein, His Tag,Human CD46, His Tag (CD6-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Cys 35 - Asp 328 (Accession AAH30594.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD46 Protein, His Tag,Human CD46, His Tag (CD6-H5226) is expressed from human 293 cells (HEK293). It contai ns AA Cys 35 - Asp 328 (Accession AAH30594.1).This protein carries a polyhistidine tag at the C-terminus. The protei 1 * 1 mg ACRBCD6-H5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou