Recombinat Protein,Human MCAM CD146 Protein, His Tag,Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession NP006491.2).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human MCAM CD146 Protein, His Tag,Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession NP006491.2).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human MCAM CD146 Protein, His Tag,Human MCAM, His Tag (CD6-H5229) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession NP006491.2).This protein carries a polyhistidine tag at the C-terminus. The ca 1 * 1 mg ACRBCD6-H5229-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou