Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The p a 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The p a 1 * 1 mg ACRBCD6-H52H9-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou