Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The p a 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The p a 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The p a 1 * 100 µG ACRBCD6-H52H9100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou