Mouse B7-2, His Tag (CD6-M52H0) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession NP062261.3). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Mouse B7-2, His Tag (CD6-M52H0) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession NP062261.3). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Mouse B7-2, His Tag (CD6-M52H0) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Glu 245 (Accession NP062261.3). 1 * 100 µG ACRBCD6-M52H0100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou