Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession F1P6D7-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession F1P6D7-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Canine CD47, His Tag (CD7-C52H5) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Asn 140 (Accession F1P6D7-1). 1 * 1 mg ACRBCD7-C52H5-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou