Recombinat Protein,Human EMMPRIN CD147 Protein, His Tag,Human EMMPRIN, His Tag (CD7-H5222) is expressed from human 293 cells (HEK29 3). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human EMMPRIN CD147 Protein, His Tag,Human EMMPRIN, His Tag (CD7-H5222) is expressed from human 293 cells (HEK29 3). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human EMMPRIN CD147 Protein, His Tag,Human EMMPRIN, His Tag (CD7-H5222) is expressed from human 293 cells (HEK29 3). It contains AA Ala 22 - His 205 (Accession NP940991.1).This protein carries a polyhistidine tag at the C-terminus 1 * 1 mg ACRBCD7-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou