Recombinat Protein,Human CD37 Protein, Fc Tag,Human CD37, Fc Tag (CD7-H526x) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Apn 240 (Accession AAI06753).This protein carries a human IgG1 Fc tag at the N-terminus. The protein h 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD37 Protein, Fc Tag,Human CD37, Fc Tag (CD7-H526x) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Apn 240 (Accession AAI06753).This protein carries a human IgG1 Fc tag at the N-terminus. The protein h 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD37 Protein, Fc Tag,Human CD37, Fc Tag (CD7-H526x) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Apn 240 (Accession AAI06753).This protein carries a human IgG1 Fc tag at the N-terminus. The protein h 1 * 1 mg ACRBCD7-H526X-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou