Recombinat Protein,Biotinylated Human EMMPRIN CD147 Protein, Avitag,His Tag,Biotinylated Human EMMPRIN, Avitag,His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession P35613-2).This protein carrie A 1 * 25 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human EMMPRIN CD147 Protein, Avitag,His Tag,Biotinylated Human EMMPRIN, Avitag,His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession P35613-2).This protein carrie A 1 * 25 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Biotinylated Human EMMPRIN CD147 Protein, Avitag™,His Tag,Biotinylated Human EMMPRIN, Avitag,His Tag (CD7-H82E0) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession P35613-2).This protein carrie A 1 * 25 µG ACRBCD7-H82E0-25UG 1 * 25 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou