Mouse CD47, His Tag (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse CD47, His Tag (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse CD47, His Tag (CD7-M522b) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735-1). 1 * 1 mg ACRBCD7-M522B-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou