Recombinat Protein,Mouse CD47 Protein, Fc Tag,Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Mouse CD47 Protein, Fc Tag,Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse CD47 Protein, Fc Tag,Mouse CD47, Fc Tag (CD7-M5251) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Lys 140 (Accession Q61735).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has o 1 * 100 µG ACRBCD7-M5251100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou