Recombinat Protein,Cynomolgus CD38 Protein, His Tag,Cynomolgus CD38, His Tag (CD8-C5223) is expressed from human 293 cells (HEK293). It contains AA Leu 44 - Ile 301 (Accession Q5VAN0-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus CD38 Protein, His Tag,Cynomolgus CD38, His Tag (CD8-C5223) is expressed from human 293 cells (HEK293). It contains AA Leu 44 - Ile 301 (Accession Q5VAN0-1).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 µG ACRBCD8-C5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou