Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 1 mg ACRBCD8-H5224-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou