Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 50 G
Fournisseur: ACROBIOSYSTEMS
Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 50 G
COMMANDE
Développer tout / Réduire tout
Human CD38, His Tag (CD8-H5224) is expressed from human 293 cells (HEK293). It contains AA Val 43 - Ile 300 (Accession NP001766). 1 * 50 µG ACRBCD8-H5224-50UG 1 * 50 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou