Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630 (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630 (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD98 Protein, His Tag,Human CD98, His Tag (CD8-H5247) is expressed from human 293 cells (HEK293). It contai ns AA Arg 206 - Ala 630  (Accession AAH01061).This protein carries a polyhistidine tag at the N-terminus. The prote a 1 * 1 mg ACRBCD8-H5247-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou