Human Cynomolgus Rhesus macaque CD28, Fc Tag (HPLC-verified) (CD8-H525a) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhe D 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Human Cynomolgus Rhesus macaque CD28, Fc Tag (HPLC-verified) (CD8-H525a) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhe D 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Human Cynomolgus Rhesus macaque CD28, Fc Tag (HPLC-verified) (CD8-H525a) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession P10747-1). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhe D 1 * 100 µG ACRBCD8-H525A100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou