Human CD16a (V176), His Tag (SPR BLI verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Human CD16a (V176), His Tag (SPR BLI verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Human CD16a (V176), His Tag (SPR  BLI verified) (CD8-H52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession P08637-1). 1 * 1 mg ACRBCD8-H52H4-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou