Biotinylated Human Cynomolgus Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhes 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Biotinylated Human Cynomolgus Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhes 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Biotinylated Human Cynomolgus Rhesus macaque CD28, Fc,Avitag (CD8-H82F2) is expressed from human 293 cells (HEK293). It contains A A Asn 19 - Pro 152 (Accession AAH93698). In the region Asn 19 - Pro 152, the AA sequence of Human, Cynomolgus and Rhes 1 * 200 µG ACRBCD8-H82F2200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou