Recombinat Protein,Mouse CD38 Protein, His Tag,Mouse CD38, His Tag (CD8-M5223) is expressed from human 293 cells (HEK293). It contai ns AA Leu 45 - Thr 304 (Accession P56528-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Mouse CD38 Protein, His Tag,Mouse CD38, His Tag (CD8-M5223) is expressed from human 293 cells (HEK293). It contai ns AA Leu 45 - Thr 304 (Accession P56528-1).This protein carries a polyhistidine tag at the C-terminus. The protein d 1 * 100 µG ACRBCD8-M5223100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou