Recombinat Protein,Human CD9 Protein, His Tag,Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession P21926-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 20 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD9 Protein, His Tag,Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession P21926-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 20 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD9 Protein, His Tag,Human CD9, His Tag (CD9-H5220) is expressed from human 293 cells (HEK293). It contains AA Ser 112 - Ile 195 (Accession P21926-1).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 20 µG ACRBCD9-H5220-20UG 1 * 20 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou