Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD99 MIC2 Protein, His Tag,Human CD99, His Tag (CD9-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 122 (Accession AAH02584).This protein carries a polyhistidine tag at the N-terminus. The prot 1 * 100 µG ACRBCD9-H5221100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou