Recombinat Protein,Human CD69 CLEC2C Protein, His Tag,Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). I t contains AA Ser 62 - Lys 199 (Accession AAH07037).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD69 CLEC2C Protein, His Tag,Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). I t contains AA Ser 62 - Lys 199 (Accession AAH07037).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD69 CLEC2C Protein, His Tag,Human CD69, His Tag (CD9-H5222) is expressed from human 293 cells (HEK293). I t contains AA Ser 62 - Lys 199 (Accession AAH07037).This protein carries a polyhistidine tag at the C-terminus. The p 1 * 1 mg ACRBCD9-H5222-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou