Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293) . It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293) . It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293) . It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 1 mg ACRBCD9-H5246-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou