Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293) . It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 100 µG
Fournisseur: ACROBIOSYSTEMS INC
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD209 DC-SIGN Protein, His Tag,Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293) . It contains AA Gln 59 - Ala 404 (Accession Q9NNX6-1).This protein carries a polyhistidine tag at the N-terminus. Th 1 * 100 µG ACRBCD9-H5246100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou