Human CD19 (20-291), His Tag (CD9-H52H2) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession P15 391-1). 1 * 500 G
Fournisseur: ACROBIOSYSTEMS
Human CD19 (20-291), His Tag (CD9-H52H2) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession P15 391-1). 1 * 500 G
COMMANDE
Développer tout / Réduire tout
Human CD19 (20-291), His Tag (CD9-H52H2) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 291 (Accession P15 391-1). 1 * 500 µG ACRBCD9-H52H2500UG 1 * 500 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou