Mouse CD16, His Tag (SPR BLI verified) (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Acc ession Q5D5I8). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Mouse CD16, His Tag (SPR BLI verified) (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Acc ession Q5D5I8). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Mouse CD16, His Tag (SPR BLI verified) (CDA-M52H8) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Acc ession Q5D5I8). 1 * 1 mg ACRBCDA-M52H8-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou