Recombinat Protein,Human CD79B Protein, Fc Tag,Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Ala 29 - Asp 159 (Accession NP000617).This protein carries a human IgG1 Fc tag at the C-terminus. The protein d 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD79B Protein, Fc Tag,Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Ala 29 - Asp 159 (Accession NP000617).This protein carries a human IgG1 Fc tag at the C-terminus. The protein d 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD79B Protein, Fc Tag,Human CD79B, Fc Tag (CDB-H5259) is expressed from human 293 cells (HEK293). It contai ns AA Ala 29 - Asp 159 (Accession NP000617).This protein carries a human IgG1 Fc tag at the C-terminus. The protein d 1 * 100 µG ACRBCDB-H5259100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou