Recombinat Protein,Human CD300c LMIR2 Protein, His Tag,Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293) . It contains AA Gly 21 - Arg 183 (Accession AAH22279).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human CD300c LMIR2 Protein, His Tag,Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293) . It contains AA Gly 21 - Arg 183 (Accession AAH22279).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human CD300c LMIR2 Protein, His Tag,Human CD300c, His Tag (CDC-H5224) is expressed from human 293 cells (HEK293) . It contains AA Gly 21 - Arg 183 (Accession AAH22279).This protein carries a polyhistidine tag at the C-terminus. Th 1 * 100 µG ACRBCDC-H5224100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou