Recombinat Protein,Cynomolgus CD3 delta Protein, His Tag,Cynomolgus CD3 delta, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession Q95LI8).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Cynomolgus CD3 delta Protein, His Tag,Cynomolgus CD3 delta, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession Q95LI8).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Cynomolgus CD3 delta Protein, His Tag,Cynomolgus CD3 delta, His Tag (CDD-C52H8) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Ala 105 (Accession Q95LI8).This protein carries a polyhistidine tag at the C-term i 1 * 1 mg ACRBCDD-C52H8-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou