Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 1 mg
Fournisseur: ACROBIOSYSTEMS
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 1 mg
COMMANDE
Développer tout / Réduire tout
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 1 mg ACRBCDE-C5226-1MG 1 * 1 mg
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou