Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 100 G
Fournisseur: ACROBIOSYSTEMS
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 100 G
COMMANDE
Développer tout / Réduire tout
Cynomolgus CD3 epsilon, His Tag (CDE-C5226) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asp 117 (Accession Q95LI5-1). 1 * 100 µG ACRBCDE-C5226100UG 1 * 100 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou