Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 200 G
Fournisseur: ACROBIOSYSTEMS
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 200 G
COMMANDE
Développer tout / Réduire tout
Recombinat Protein,Human E-Selectin CD62E Protein, His Tag,Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession NP000441.2).This protein carries a polyhistidine tag at the C-ter 1 * 200 µG ACRBCDE-H5222200UG 1 * 200 µG
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou